Learn More
Med Vet International Miltex Brown-Adson Tissue Forceps, 4.75in, Tungsten Carbide, 7 X 7 Grasping Teeth, Cross Serrated Tying Platform, German, 6-124Tc
Catalog No. 502830052
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Brown-Adson Tissue Forceps. Adson Tissue Forceps are for holding and manipulating delicate tissues. Adson tissue forceps generally have a wide thumb grasp area with short jaws that have narrow tips. They are made from premium-grade German stainless steel. Specifications: 4.75in, Tungsten Carbide, 7 X 7 Grasping Teeth, Cross Serrated Tying Platform, 6-124TC
Catalog No. 50-283-0052 Supplier Med Vet International Supplier No. MIL6124TC
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Brown-Adson Tissue Forceps. Adson Tissue Forceps are for holding and manipulating delicate tissues. Adson tissue forceps generally have a wide thumb grasp area with short jaws that have narrow tips. They are made from premium-grade German stainless steel. Specifications: 4.75in, Tungsten Carbide, 7 X 7 Grasping Teeth, Cross Serrated Tying Platform, 6-124TC