Learn More
Med Vet International Miltex Carmalt Splinter Forceps, 4-1/8in, Straight, Serrated, German, 6-308
Catalog No. 502828980
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Carmalt Splinter Forceps. Carmalt Splinter forceps are designed explicitly with serrated, pointed tips and a broad grasping surface that assures a steady and secure grip of the particles that need to be removed. They are made from premium-grade German stainless steel. Specifications: 4-1/8in, Straight, Serrated, 6-308
Catalog No. 50-282-8980 Supplier Med Vet International Supplier No. MIL6308
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Carmalt Splinter Forceps. Carmalt Splinter forceps are designed explicitly with serrated, pointed tips and a broad grasping surface that assures a steady and secure grip of the particles that need to be removed. They are made from premium-grade German stainless steel. Specifications: 4-1/8in, Straight, Serrated, 6-308