Learn More
Med Vet International Miltex Collin Forceps, 7.75in, With 1-1/8in Wide Jaws, German, 16-54
Catalog No. 502832467
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Collin Forceps. Collin Forceps are used for Thoracic and Cardiovascular procedures. They are made from premium-grade German stainless steel. Specifications: 7.75in, With 1-1/8in Wide Jaws, 16-54
Catalog No. 50-283-2467 Supplier Med Vet International Supplier No. MIL1654
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Collin Forceps. Collin Forceps are used for Thoracic and Cardiovascular procedures. They are made from premium-grade German stainless steel. Specifications: 7.75in, With 1-1/8in Wide Jaws, 16-54