Learn More
Med Vet International Miltex Cushing Dressing Forceps, 6.75in, Serrated, Scraper End, German, 6-188
Catalog No. 502830394
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Cushing Dressing Forceps. The Cushing Dressing Forceps is a thumb-type, clamping device that offers a wide range of benefits. They are made from premium-grade German stainless steel. Specifications: 6.75in, Serrated, Scraper End, 6-188
Catalog No. 50-283-0394 Supplier Med Vet International Supplier No. MIL6188
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Cushing Dressing Forceps. The Cushing Dressing Forceps is a thumb-type, clamping device that offers a wide range of benefits. They are made from premium-grade German stainless steel. Specifications: 6.75in, Serrated, Scraper End, 6-188