Learn More
Med Vet International Miltex Debakey-Diethrich Atraumatic Vascular Tissue Forceps, 8in., 1.5mm Wide Tips
Catalog No. 502829536
Encompass_Preferred
The Miltex 24-572 DeBakey-Diethrich atraumatic vascular tissue forceps are made of OR grade German stainless steel and measure 8" in length. They have a non-locking thumb handle and straight tips that are 1.5mm wide and serrated.
Catalog No. 50-282-9536 Supplier Med Vet International Supplier No. MIL24572
The Miltex 24-572 DeBakey-Diethrich atraumatic vascular tissue forceps are made of OR grade German stainless steel and measure 8" in length. They have a non-locking thumb handle and straight tips that are 1.5mm wide and serrated.