Learn More
Med Vet International Miltex Debakey Tissue Forceps, 9.75in, Tips 2.5mm Wide, German, 24-554
Catalog No. 502829458
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Debakey Tissue Forceps. Debakey Thoracic Tissue Forceps are used for grasping and manipulating tissues in cardiovascular and thoracic procedures. They are made from premium-grade German stainless steel. Specifications: 9.75in, Tips 2.5mm Wide, 24-554
Catalog No. 50-282-9458 Supplier Med Vet International Supplier No. MIL24554
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Debakey Tissue Forceps. Debakey Thoracic Tissue Forceps are used for grasping and manipulating tissues in cardiovascular and thoracic procedures. They are made from premium-grade German stainless steel. Specifications: 9.75in, Tips 2.5mm Wide, 24-554