Learn More
Med Vet International Miltex Doyen intestinal Hemostatic Forceps, 9in., Straight, Longitudinal Serrations
Catalog No. 502832472
Encompass_Preferred
The Miltex 16-160 Doyen intestinal forceps are made of OR grade German stainless steel and measure 9" in length. They have a finger ring handle with a ratchet lock and straight, flexible blades with longitudinal serrations
Catalog No. 50-283-2472 Supplier Med Vet International Supplier No. MIL16160
The Miltex 16-160 Doyen intestinal forceps are made of OR grade German stainless steel and measure 9" in length. They have a finger ring handle with a ratchet lock and straight, flexible blades with longitudinal serrations