Learn More
Med Vet International Miltex Dressing Forceps, 10in, Standard Pattern, Serrated Handles, Serrated Tips, German, 6-16
Catalog No. 502830392
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Dressing Forceps. Dressing forceps are used when dressing wounds. They hold gauze and other dressings. They may also be used during wound debridement to remove infected or necrotic tissue or debris from the wound. They are made from premium-grade German stainless steel. Specifications: 10in, Standard Pattern, Serrated Handles, Serrated Tips, 6-16
Catalog No. 50-283-0392 Supplier Med Vet International Supplier No. MIL616
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Dressing Forceps. Dressing forceps are used when dressing wounds. They hold gauze and other dressings. They may also be used during wound debridement to remove infected or necrotic tissue or debris from the wound. They are made from premium-grade German stainless steel. Specifications: 10in, Standard Pattern, Serrated Handles, Serrated Tips, 6-16