Learn More
Med Vet International Miltex Meisterhand Eye Dressing Forceps, 4in, Standard, Half Curved, Serrated, 0.8mm Wide At Tip, German, Mh18-782
Catalog No. 502831553
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Eye Dressing Forceps. Eye Dressing Forceps are used in ophthalmic procedures for grasping sterile equipment and removing tissues. They are made from premium-grade German stainless steel. Specifications: 4in, Standard, Half Curved, Serrated, 0.8mm Wide At Tip, MH18-782
Catalog No. 50-283-1553 Supplier Med Vet International Supplier No. MILMH18782
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Eye Dressing Forceps. Eye Dressing Forceps are used in ophthalmic procedures for grasping sterile equipment and removing tissues. They are made from premium-grade German stainless steel. Specifications: 4in, Standard, Half Curved, Serrated, 0.8mm Wide At Tip, MH18-782