Learn More
Med Vet International Miltex Ferris-Smith Tissue Forceps, 6.75in, 2 X 3 Teeth, German, 26-958
Catalog No. 502832287
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Ferris-Smith Tissue Forceps. Ferris Smith Tissue Forceps are heavyweight forceps used with fibrous tissue in colorectal procedures. They are made from premium-grade German stainless steel. Specifications: 6.75in, 2 X 3 Teeth, 26-958
Catalog No. 50-283-2287 Supplier Med Vet International Supplier No. MIL26958
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Ferris-Smith Tissue Forceps. Ferris Smith Tissue Forceps are heavyweight forceps used with fibrous tissue in colorectal procedures. They are made from premium-grade German stainless steel. Specifications: 6.75in, 2 X 3 Teeth, 26-958