Learn More
Med Vet International Miltex Ferguson Angiotribe Hemostatic Forceps, 7.5in., Curved
Catalog No. 502832389
Encompass_Preferred
The Miltex 7-266 Ferguson angiotribe hemostatic forceps are made of OR grade German stainless steel and measure 7.5" in length. They have a finger ring handle with a ratchet lock and curved jaws with serrations.
Catalog No. 50-283-2389 Supplier Med Vet International Supplier No. MIL7266
The Miltex 7-266 Ferguson angiotribe hemostatic forceps are made of OR grade German stainless steel and measure 7.5" in length. They have a finger ring handle with a ratchet lock and curved jaws with serrations.