Learn More
Med Vet International Miltex Ferguson Angiotribe Hemostat Forceps, 6.25in, Curved, German, 7-262
Catalog No. 502832388
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Ferguson Angiotribe Hemostat Forceps. Ferguson Angiotribe Forceps are used for hemostasis and prevention of subsequent bleeding. They are made from premium-grade German stainless steel. Specifications: 6.25in, Curved, 7-262
Catalog No. 50-283-2388 Supplier Med Vet International Supplier No. MIL7262
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Ferguson Angiotribe Hemostat Forceps. Ferguson Angiotribe Forceps are used for hemostasis and prevention of subsequent bleeding. They are made from premium-grade German stainless steel. Specifications: 6.25in, Curved, 7-262