Learn More
Med Vet International Miltex Foerster Sponge Forceps, 9.75in, Curved, Smooth Jaws, German, 7-612
Supplier:  Med Vet International MIL7612
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Foerster Sponge Forceps. Foerster Sponge Forceps are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site or hemostasis. They are made from premium-grade German stainless steel. Specifications: 9.75in, Curved, Smooth Jaws, 7-612
Catalog No. 50-282-9099