Learn More
Med Vet International Miltex Foerster Eye Dressing Forceps, 3.75in Octagonal Grip, Full Curved, Serrated, 0.5mm Wide, German, 18-795A
Catalog No. 502830527
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Foerster Eye Dressing Forceps. Eye Dressing Forceps are used in ophthalmic procedures for grasping sterile equipment and removing tissues. They are made from premium-grade German stainless steel. Specifications: 3.75in Octagonal Grip, Full Curved, Serrated, 0.5mm Wide, 18-795A
Catalog No. 50-283-0527 Supplier Med Vet International Supplier No. MIL18795A
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Foerster Eye Dressing Forceps. Eye Dressing Forceps are used in ophthalmic procedures for grasping sterile equipment and removing tissues. They are made from premium-grade German stainless steel. Specifications: 3.75in Octagonal Grip, Full Curved, Serrated, 0.5mm Wide, 18-795A