Learn More
Med Vet International Miltex Meisterhand Gemini-Mixter Hemostatic Forceps, 5.5in, Delicate, Full Curved Jaws, German, Mh7-255
Catalog No. 502832164
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Gemini-Mixter Hemostatic Forceps. Gemini-Mixter Forceps are comprised with fully curved delicate and serrated jaws that allow surgeons to get a hold of blood vessels that may be obstructed by tissue. They are made from premium-grade German stainless steel. Specifications: 5.5in, Delicate, Full Curved Jaws, MH7-255
Catalog No. 50-283-2164 Supplier Med Vet International Supplier No. MILMH7255
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Gemini-Mixter Hemostatic Forceps. Gemini-Mixter Forceps are comprised with fully curved delicate and serrated jaws that allow surgeons to get a hold of blood vessels that may be obstructed by tissue. They are made from premium-grade German stainless steel. Specifications: 5.5in, Delicate, Full Curved Jaws, MH7-255