Learn More
Med Vet International Miltex Gerald Dressing Forceps, 7in, Curved, Serrated, German, 6-184
Catalog No. 502830393
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Gerald Dressing Forceps. Gerald Dressing Forceps are used for removing and applying dressings. These forceps are also widely used to remove foreign body particles and ear tube particles. They are made from premium-grade German stainless steel. Specifications: 7in, Curved, Serrated, 6-184
Catalog No. 50-283-0393 Supplier Med Vet International Supplier No. MIL6184
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Gerald Dressing Forceps. Gerald Dressing Forceps are used for removing and applying dressings. These forceps are also widely used to remove foreign body particles and ear tube particles. They are made from premium-grade German stainless steel. Specifications: 7in, Curved, Serrated, 6-184