Learn More
Med Vet International Miltex Meisterhand Gerald Dressing Forceps, 7in, Straight, Serrated, German, Mh6-180
Catalog No. 502832343
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Gerald Dressing Forceps. Gerald Dressing Forceps are used for removing and applying dressings. These forceps are also widely used to remove foreign body particles and ear tube particles. They are made from premium-grade German stainless steel. Specifications: 7in, Straight, Serrated, MH6-180
Catalog No. 50-283-2343 Supplier Med Vet International Supplier No. MILMH6180
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Gerald Dressing Forceps. Gerald Dressing Forceps are used for removing and applying dressings. These forceps are also widely used to remove foreign body particles and ear tube particles. They are made from premium-grade German stainless steel. Specifications: 7in, Straight, Serrated, MH6-180