Learn More
Med Vet International Miltex Gill-Hess Irish Forceps, 3in, Bayonet-Shaped, With 1X2 Teeth Forward, 0.5mm Wide Tip
Catalog No. 502831771
Encompass_Preferred
The Miltex 18-830 Gill-Hess Irish forceps are made of OR grade German stainless steel and measure 3" in length. They have a non-locking thumb handle and are bayonet-shaped. The tips are 0.5mm wide with 1 x 2 teeth forward.
Catalog No. 50-283-1771 Supplier Med Vet International Supplier No. MIL18830
The Miltex 18-830 Gill-Hess Irish forceps are made of OR grade German stainless steel and measure 3" in length. They have a non-locking thumb handle and are bayonet-shaped. The tips are 0.5mm wide with 1 x 2 teeth forward.