Learn More
Med Vet International Miltex Halsted Mosquito Forceps, 4.5in, Jaws Angled 45 Degrees On Flat, Extra Delicate, German, 7-20
Catalog No. 502831980
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Halsted Mosquito Forceps. Halsted-Mosquito hemostatic forceps are versatile, ratcheted, finger ring forceps used for clamping vessels and controlling blow flow and other clamping tasks in general surgical procedures. They are made from premium-grade German stainless steel. Specifications: 4.5in, Jaws Angled 45 Degrees On Flat, Extra Delicate, 7-20
Catalog No. 50-283-1980 Supplier Med Vet International Supplier No. MIL720
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Halsted Mosquito Forceps. Halsted-Mosquito hemostatic forceps are versatile, ratcheted, finger ring forceps used for clamping vessels and controlling blow flow and other clamping tasks in general surgical procedures. They are made from premium-grade German stainless steel. Specifications: 4.5in, Jaws Angled 45 Degrees On Flat, Extra Delicate, 7-20