Learn More
Med Vet International Miltex House Ear Forceps, 3-1/8in Shaft, 4.5mm Finely Serrated Jaws, Extra Delicate, Alligator Type, German, 19-415
Catalog No. 502829238
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex House Ear Forceps. Ear Forceps can be used for a variety of ENT procedures. They are made from premium-grade German stainless steel. Specifications: 3-1/8in Shaft, 4.5mm Finely Serrated Jaws, Extra Delicate, Alligator Type, 19-415
Catalog No. 50-282-9238 Supplier Med Vet International Supplier No. MIL19415
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex House Ear Forceps. Ear Forceps can be used for a variety of ENT procedures. They are made from premium-grade German stainless steel. Specifications: 3-1/8in Shaft, 4.5mm Finely Serrated Jaws, Extra Delicate, Alligator Type, 19-415