Learn More
Med Vet International Miltex Hudson (Ewald) Tissue Forceps, 4.75in, 1 X 2 Teeth, German, 6-114
Catalog No. 502830419
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Hudson (Ewald) Tissue Forceps. Hudson Tissue Forceps are used to hold the soft tissues during multiple surgical procedures. They are made from premium-grade German stainless steel. Specifications: 4.75in, 1 X 2 Teeth, 6-114
Catalog No. 50-283-0419 Supplier Med Vet International Supplier No. MIL6114
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Hudson (Ewald) Tissue Forceps. Hudson Tissue Forceps are used to hold the soft tissues during multiple surgical procedures. They are made from premium-grade German stainless steel. Specifications: 4.75in, 1 X 2 Teeth, 6-114