Learn More
Fine Science Tools FORCEP INSECT COLLECTING
SDP
Catalog No. NC9450838
Encompass
Tip Shape: Straight/ Tips: Sharp/ Tip Dimensions: 0.9 x 0.15mm/ Alloy / Material: Stainless Steel/ Length: 10cm
Catalog No. NC9450838 Supplier Fine Science Tools Supplier No. 2602910
Tip Shape: Straight/ Tips: Sharp/ Tip Dimensions: 0.9 x 0.15mm/ Alloy / Material: Stainless Steel/ Length: 10cm