Learn More
Med Vet International Miltex Jacobson Mosquito Micro Forceps, 4.75in, Curved, Extra Delicate, German, 17-2602
Catalog No. 502830372
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Jacobson Mosquito Micro Forceps. Jacobson Micro Mosquito Forceps are used as hemostats to control the bleeding from blood vessels. They are made from premium-grade German stainless steel. Specifications: 4.75in, Curved, Extra Delicate, 17-2602
Catalog No. 50-283-0372 Supplier Med Vet International Supplier No. MIL172602
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Jacobson Mosquito Micro Forceps. Jacobson Micro Mosquito Forceps are used as hemostats to control the bleeding from blood vessels. They are made from premium-grade German stainless steel. Specifications: 4.75in, Curved, Extra Delicate, 17-2602