Learn More
Med Vet International Miltex Jeweler-Style Forceps, 4.5in, Non-Magnetic, Stainless Steel, Style 7, Fine Jaw, Curved, 17-307
Catalog No. 502830429
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Jeweler-Style Forceps. Jeweler's Forceps are highly versatile and known as handheld surgical instruments and are used to lift or grasp tissues during surgical procedures. They are made from premium-grade surgical stainless steel. Specifications: 4.5in, Non-Magnetic, Stainless Steel, Style 7, Fine Jaw, Curved, 17-307
Catalog No. 50-283-0429 Supplier Med Vet International Supplier No. MIL17307
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Jeweler-Style Forceps. Jeweler's Forceps are highly versatile and known as handheld surgical instruments and are used to lift or grasp tissues during surgical procedures. They are made from premium-grade surgical stainless steel. Specifications: 4.5in, Non-Magnetic, Stainless Steel, Style 7, Fine Jaw, Curved, 17-307