Learn More
Med Vet International Miltex Kelly Tissue Forceps, 9.75in, 1 X 2 Teeth, Slotted Gutch Handles, German, 6-200
Catalog No. 502828979
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Kelly Tissue Forceps. Kelly Tissue Forceps are well-suited for demand in cardiovascular procedures. They are made from premium-grade German stainless steel. Specifications: 9.75in, 1 X 2 Teeth, Slotted Gutch Handles, 6-200
Catalog No. 50-282-8979 Supplier Med Vet International Supplier No. MIL6200
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Kelly Tissue Forceps. Kelly Tissue Forceps are well-suited for demand in cardiovascular procedures. They are made from premium-grade German stainless steel. Specifications: 9.75in, 1 X 2 Teeth, Slotted Gutch Handles, 6-200