Learn More
Med Vet International Miltex Kevorkian-Younge Uterine Biopsy Forceps, 9.75in Shaft, 3.2mm X 10mm Bite, German, 30-1482
Catalog No. 502832113
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Kevorkian-Younge Uterine Biopsy Forceps. Kevorkian-Younge Biopsy Forceps are designed to take a tissue sample from the uterus. They are made from premium-grade German stainless steel. Specifications: 9.75in Shaft, 3.2mm X 10mm Bite, 30-1482
Catalog No. 50-283-2113 Supplier Med Vet International Supplier No. MIL301482
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Kevorkian-Younge Uterine Biopsy Forceps. Kevorkian-Younge Biopsy Forceps are designed to take a tissue sample from the uterus. They are made from premium-grade German stainless steel. Specifications: 9.75in Shaft, 3.2mm X 10mm Bite, 30-1482