Learn More
Med Vet International Miltex Lahey Gall Duct Forceps, 9in, Curved, German, 14-49
Catalog No. 502831270
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Lahey Gall Duct Forceps. Lahey Gall Duct Forceps are highly versatile surgical instruments used for dissections in those body areas that may not be viewed easily. They are made from premium-grade German stainless steel. Specifications: 9in, Curved, 14-49
Catalog No. 50-283-1270 Supplier Med Vet International Supplier No. MIL1449
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Lahey Gall Duct Forceps. Lahey Gall Duct Forceps are highly versatile surgical instruments used for dissections in those body areas that may not be viewed easily. They are made from premium-grade German stainless steel. Specifications: 9in, Curved, 14-49