Learn More
Med Vet International Miltex Mccullough Utility Forceps, 4in, 1 X 2 Teeth, Cross-Serrated, 1.5mm Wide At Tip, German, 18-966
Catalog No. 502831814
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Mccullough Utility Forceps. Mccullough Utility Forceps are a general purpose tool used in many ophthalmologic surgeries. They are made from premium-grade German stainless steel. Specifications: 4in, 1 X 2 Teeth, Cross-Serrated, 1.5mm Wide At Tip, 18-966
Catalog No. 50-283-1814 Supplier Med Vet International Supplier No. MIL18966
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Mccullough Utility Forceps. Mccullough Utility Forceps are a general purpose tool used in many ophthalmologic surgeries. They are made from premium-grade German stainless steel. Specifications: 4in, 1 X 2 Teeth, Cross-Serrated, 1.5mm Wide At Tip, 18-966