Learn More
Med Vet International Miltex Meisterhand Mcpherson Suture Tying Forceps, 3.5in, Angled, Smooth Platform 0.3 X 5 mm, No Teeth, German, Mh18-949
Supplier:  Med Vet International MILMH18949
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Mcpherson Suture Tying Forceps. Unlike regular forceps that meet at their tips when they are closed, these suture tying forceps with tying platforms close flat along a long surface from the tip. This feature allows for an easy and reliable grip of surgical suture materials. They are made from premium-grade German stainless steel. Specifications: 3.5in, Angled, Smooth Platform 0.3 X 5 mm, No Teeth, MH18-949
Catalog No. 50-283-1156