Learn More
Electron Microscopy Sciences Micro Dissecting Forceps Membrane Dissector with Curved Blades
SDP
Catalog No. 5034615
Encompass_Preferred
20 gauge stainless steel, with 3 mm tips.
Blades: Curved
Catalog No. 50-346-15 Supplier Electron Microscopy Sciences Supplier No. 7793204
20 gauge stainless steel, with 3 mm tips.
Blades: Curved