Learn More
Med Vet International Miltex Michel Clip Applying & Removing Forceps, 4-7/8in, Double-Ended, German, 9-60
Catalog No. 502832096
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Michel Clip Applying & Removing Forceps. Clip Applying Forceps allow for a secure grasp of suture clips. They are made from premium-grade German stainless steel. Specifications: 4-7/8in, Double-Ended, 9-60
Catalog No. 50-283-2096 Supplier Med Vet International Supplier No. MIL960
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Michel Clip Applying & Removing Forceps. Clip Applying Forceps allow for a secure grasp of suture clips. They are made from premium-grade German stainless steel. Specifications: 4-7/8in, Double-Ended, 9-60