Learn More
Med Vet International Miltex Micro Adson Dressing Forceps, 4.75in., With 1X2 Teeth, 0.5mm Wide Tips
Catalog No. 502830370
Encompass_Preferred
The Miltex 17-2510 micro Adson dressing forceps are made of OR grade German stainless steel and measure 4.75" in length. They have a non-locking thumb handle and straight tips that are 0.5mm wide, serrated, and feature 1 x 2 teeth.
Catalog No. 50-283-0370 Supplier Med Vet International Supplier No. MIL172510
 Restricted Item
Add to Cart
Add to Cart
The Miltex 17-2510 micro Adson dressing forceps are made of OR grade German stainless steel and measure 4.75" in length. They have a non-locking thumb handle and straight tips that are 0.5mm wide, serrated, and feature 1 x 2 teeth.