Learn More
Med Vet International Miltex Hartman Mosquito Hemostatic Forceps, 3.5in., Curved
Catalog No. 502832386
Encompass_Preferred
The Miltex 7-26 Hartman mosquito forceps are made of OR grade German stainless steel and measure 3.5" in length. They have a finger ring handle with a ratchet lock and curved tips that are serrated.
Catalog No. 50-283-2386 Supplier Med Vet International Supplier No. MIL726
The Miltex 7-26 Hartman mosquito forceps are made of OR grade German stainless steel and measure 3.5" in length. They have a finger ring handle with a ratchet lock and curved tips that are serrated.