Learn More
Med Vet International Miltex Mosquito Hemostatic Forceps, 7.75in, Right Angle, Delicate, German, 7-254
Catalog No. 502832385
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Mosquito Hemostatic Forceps. Mosquito forceps are a small, straight or curved hemostatic forceps used to hold delicate tissue or compress a bleeding vessel. They are made from premium-grade German stainless steel. Specifications: 7.75in, Right Angle, Delicate, 7-254
Catalog No. 50-283-2385 Supplier Med Vet International Supplier No. MIL7254
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Mosquito Hemostatic Forceps. Mosquito forceps are a small, straight or curved hemostatic forceps used to hold delicate tissue or compress a bleeding vessel. They are made from premium-grade German stainless steel. Specifications: 7.75in, Right Angle, Delicate, 7-254