Learn More
Med Vet International Miltex Noyes Nasal Dressing Forceps, 5.5in Shaft, Serrated Jaws, German, 20-140
Catalog No. 502829345
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Noyes Nasal Dressing Forceps. Noyes Nasal Dressing Forceps are used in a variety of ENT procedures. The long shaft is ideal for foreign body removal in small deep crevices. They are made from premium-grade German stainless steel. Specifications: 5.5in Shaft, Serrated Jaws, 20-140
Catalog No. 50-282-9345 Supplier Med Vet International Supplier No. MIL20140
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Noyes Nasal Dressing Forceps. Noyes Nasal Dressing Forceps are used in a variety of ENT procedures. The long shaft is ideal for foreign body removal in small deep crevices. They are made from premium-grade German stainless steel. Specifications: 5.5in Shaft, Serrated Jaws, 20-140