Learn More
Med Vet International Miltex Peet Splinter Forceps, 4.5in, Double Curve, German, 6-350
Catalog No. 502829171
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Peet Splinter Forceps. Splinter Forceps are a fine-tipped forcep used for grasping and removing small foreign objects. They are made from premium-grade German stainless steel. Specifications: 4.5in, Double Curve, 6-350
Catalog No. 50-282-9171 Supplier Med Vet International Supplier No. MIL6350
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Peet Splinter Forceps. Splinter Forceps are a fine-tipped forcep used for grasping and removing small foreign objects. They are made from premium-grade German stainless steel. Specifications: 4.5in, Double Curve, 6-350