Learn More
Med Vet International Miltex Pierse Colibri Corneal Forceps, 3in, Angled, 0.35mm Wide At Tip, German, 18-881
Catalog No. 502831811
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Pierse Colibri Corneal Forceps. The Pierse-Colibri Corneal Forceps is used to manipulate the cornea during ocular surgical procedures. They are made from premium-grade German stainless steel. Specifications: 3in, Angled, 0.35mm Wide At Tip, 18-881
Catalog No. 50-283-1811 Supplier Med Vet International Supplier No. MIL18881
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Pierse Colibri Corneal Forceps. The Pierse-Colibri Corneal Forceps is used to manipulate the cornea during ocular surgical procedures. They are made from premium-grade German stainless steel. Specifications: 3in, Angled, 0.35mm Wide At Tip, 18-881