Learn More
Med Vet International Miltex Platypus Nail Pulling Forceps, 5.5in, Narrow Jaws, 4.2mm Wide, Stainless, German, 40-235
Catalog No. 502831160
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Platypus Nail Pulling Forceps. Platypus Nail Pulling Forceps feature standard or narrow jaws used to split ingrown toenails and then remove the severed portion of the nail from the soft tissue. They are made from premium-grade German stainless steel. Specifications: 5.5in, Narrow Jaws, 4.2mm Wide, Stainless, 40-235
Catalog No. 50-283-1160 Supplier Med Vet International Supplier No. MIL40235
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Platypus Nail Pulling Forceps. Platypus Nail Pulling Forceps feature standard or narrow jaws used to split ingrown toenails and then remove the severed portion of the nail from the soft tissue. They are made from premium-grade German stainless steel. Specifications: 5.5in, Narrow Jaws, 4.2mm Wide, Stainless, 40-235