Learn More
Med Vet International Miltex Post Mortem Arterial Fixation Forceps, 7.5in, Angled, 50-582
Catalog No. 502831828
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Post Mortem Arterial Fixation Forceps. Arterial Fixation Forceps are excellent for general use. Also ideal for holding arterial tubes. They are made from premium-grade surgical stainless steel. Specifications: 7.5in, Angled, 50-582
Catalog No. 50-283-1828 Supplier Med Vet International Supplier No. MIL50582
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Post Mortem Arterial Fixation Forceps. Arterial Fixation Forceps are excellent for general use. Also ideal for holding arterial tubes. They are made from premium-grade surgical stainless steel. Specifications: 7.5in, Angled, 50-582