Learn More
Med Vet International Miltex Potts-Smith Dressing Forceps, 7.25in, Tips 1.6mm Wide, Serrated, German, 6-154
Catalog No. 502830390
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Potts-Smith Dressing Forceps. Potts-Smith Dressing Forceps are used for changing dressings, and for dissecting fine or single-layer tissue. They are made from premium-grade German stainless steel. Specifications: 7.25in, Tips 1.6mm Wide, Serrated, 6-154
Catalog No. 50-283-0390 Supplier Med Vet International Supplier No. MIL6154
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Potts-Smith Dressing Forceps. Potts-Smith Dressing Forceps are used for changing dressings, and for dissecting fine or single-layer tissue. They are made from premium-grade German stainless steel. Specifications: 7.25in, Tips 1.6mm Wide, Serrated, 6-154