Learn More
Med Vet International Miltex Schubert Uterine Biopsy Forceps, 9.75in, Angled Shanks, 5.25mm X 9.2mm Bite, German, 30-1430
Catalog No. 502832341
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Schubert Uterine Biopsy Forceps. Schubert Biopsy Forceps are designed with triangular bites to take biopsy samples from the uterine tissue. They are made from premium-grade German stainless steel. Specifications: 9.75in, Angled Shanks, 5.25mm X 9.2mm Bite, 30-1430
Catalog No. 50-283-2341 Supplier Med Vet International Supplier No. MIL301430
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Schubert Uterine Biopsy Forceps. Schubert Biopsy Forceps are designed with triangular bites to take biopsy samples from the uterine tissue. They are made from premium-grade German stainless steel. Specifications: 9.75in, Angled Shanks, 5.25mm X 9.2mm Bite, 30-1430