Learn More
Med Vet International Miltex Meisterhand Schnidt-Sawtell Hemostatic Forceps, 7.5in, Full Curved, Closed Ring Handles, German, Mh22-668
Catalog No. 502829714
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Schnidt-Sawtell Hemostatic Forceps. Schnidt-Sawtell Forceps are hemostatic tonsil forceps. They are made from premium-grade German stainless steel. Specifications: 7.5in, Full Curved, Closed Ring Handles, MH22-668
Catalog No. 50-282-9714 Supplier Med Vet International Supplier No. MILMH22668
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Schnidt-Sawtell Hemostatic Forceps. Schnidt-Sawtell Forceps are hemostatic tonsil forceps. They are made from premium-grade German stainless steel. Specifications: 7.5in, Full Curved, Closed Ring Handles, MH22-668