Learn More
Med Vet International Miltex Meisterhand Meisterhard Schroeder Tenaculum Forceps, 10in, Rounded Jaw With Non-Overlapping Atraumatic Tips, German, Mh30-966Atr
Catalog No. 502829771
Encompass_Preferred
The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand MeisterHand Schroeder Tenaculum Forceps. Tenaculum Forceps are commonly used in Gynecological & Obstetrical procedures involving manipulation of the cervix or uterus, most commonly D&C, and vaginal or abdominal hysterectomy. They are also used to grasp, hold, and move vessels and arteries during surgical procedures. They are made from premium-grade German stainless steel. Specifications: 10in, Rounded Jaw With Non-Overlapping Atraumatic Tips, MH30-966ATR
Catalog No. 50-282-9771 Supplier Med Vet International Supplier No. MILMH30966ATR
 Restricted Item
Add to Cart
Add to Cart
The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand MeisterHand Schroeder Tenaculum Forceps. Tenaculum Forceps are commonly used in Gynecological & Obstetrical procedures involving manipulation of the cervix or uterus, most commonly D&C, and vaginal or abdominal hysterectomy. They are also used to grasp, hold, and move vessels and arteries during surgical procedures. They are made from premium-grade German stainless steel. Specifications: 10in, Rounded Jaw With Non-Overlapping Atraumatic Tips, MH30-966ATR