Learn More
Med Vet International Miltex Schroeder Vulsellum Uterine Forceps, 9.75in, Straight, 2 X 2 Teeth, German, 30-1000
Catalog No. 502832062
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Schroeder Vulsellum Uterine Forceps. Schroeder Vulsellum Forceps are designed to grip the uterus during surgical procedures. They are made from premium-grade German stainless steel. Specifications: 9.75in, Straight, 2 X 2 Teeth, 30-1000
Catalog No. 50-283-2062 Supplier Med Vet International Supplier No. MIL301000
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Schroeder Vulsellum Uterine Forceps. Schroeder Vulsellum Forceps are designed to grip the uterus during surgical procedures. They are made from premium-grade German stainless steel. Specifications: 9.75in, Straight, 2 X 2 Teeth, 30-1000