Learn More
Med Vet International Miltex Semken Tissue Forceps, 5in, 1 X 2 Teeth, German, 6-106
Catalog No. 502830426
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Semken Tissue Forceps. Semken Tissue (Thumb) Forceps are used for holding and manipulating delicate tissues. They are made from premium-grade German stainless steel. Specifications: 5in, 1 X 2 Teeth, 6-106
Catalog No. 50-283-0426 Supplier Med Vet International Supplier No. MIL6106
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Semken Tissue Forceps. Semken Tissue (Thumb) Forceps are used for holding and manipulating delicate tissues. They are made from premium-grade German stainless steel. Specifications: 5in, 1 X 2 Teeth, 6-106