Learn More
Med Vet International Miltex Snellen Entropium Forceps, 3-7/8in, Right, Solid Plate And Ring 1-3/8in Wide, German, 18-1162
Catalog No. 502830435
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Snellen Entropium Forceps. Snellen Entropium Forceps are a commonly used tool to hold the eyelid in place during corrective surgery. They are made from premium-grade German stainless steel. Specifications: 3-7/8in, Right, Solid Plate And Ring 1-3/8in Wide, 18-1162
Catalog No. 50-283-0435 Supplier Med Vet International Supplier No. MIL181162
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Snellen Entropium Forceps. Snellen Entropium Forceps are a commonly used tool to hold the eyelid in place during corrective surgery. They are made from premium-grade German stainless steel. Specifications: 3-7/8in, Right, Solid Plate And Ring 1-3/8in Wide, 18-1162