Learn More
Med Vet International Miltex Padgett Micro Surgical Forceps, 4-3/8in, Angled, Pm-4928
Catalog No. 502832133
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Padgett Micro Surgical Forceps. Micro Surgical Forceps are used to grasp and manipulate delicate tissues in small surgical sites. They are made from premium-grade surgical stainless steel. Specifications: 4-3/8in, Angled, PM-4928
Catalog No. 50-283-2133 Supplier Med Vet International Supplier No. MILPM4928
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Padgett Micro Surgical Forceps. Micro Surgical Forceps are used to grasp and manipulate delicate tissues in small surgical sites. They are made from premium-grade surgical stainless steel. Specifications: 4-3/8in, Angled, PM-4928