Learn More
Med Vet International Miltex Meisterhand Takahashi Nasal Forceps, 4.5in Shaft, 2.5mm X 10mm Jaws, German, Mh20-570
Catalog No. 502831154
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Takahashi Nasal Forceps. Takahashi Nasal Forceps are commonly used in nasal surgical procedures. They may be used for soft tissue and cartilage removal, removal of excess bony fragments and biopsy sampling. They are made from premium-grade German stainless steel. Specifications: 4.5in Shaft, 2.5mm X 10mm Jaws, MH20-570
Catalog No. 50-283-1154 Supplier Med Vet International Supplier No. MILMH20570
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex MeisterHand Takahashi Nasal Forceps. Takahashi Nasal Forceps are commonly used in nasal surgical procedures. They may be used for soft tissue and cartilage removal, removal of excess bony fragments and biopsy sampling. They are made from premium-grade German stainless steel. Specifications: 4.5in Shaft, 2.5mm X 10mm Jaws, MH20-570