Learn More
Med Vet International Miltex Tissue Forceps, 5.5in., With 1X2 Teeth, Standard Pattern, Serrated Handles
Catalog No. 502829172
Encompass_Preferred
The Miltex 6-44 tissue forceps are made of OR grade German stainless steel and measure 5.5" in length. They have a non-locking, serrated thumb handles and the tips are straight with 1 x 2 teeth.
Catalog No. 50-282-9172 Supplier Med Vet International Supplier No. MIL644
The Miltex 6-44 tissue forceps are made of OR grade German stainless steel and measure 5.5" in length. They have a non-locking, serrated thumb handles and the tips are straight with 1 x 2 teeth.