Learn More
Med Vet International Miltex Uterine Biopsy Forceps, 9in Shaft, 6mm Bite, German, 30-1480
Catalog No. 502832441
Encompass_Preferred
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Uterine Biopsy Forceps. Uterine Biopsy Forceps are commonly used for biopsy of the uterine cervix in colposcopy. They are made from premium-grade German stainless steel. Specifications: 9in Shaft, 6mm Bite, 30-1480
Catalog No. 50-283-2441 Supplier Med Vet International Supplier No. MIL301480
The Miltex name is synonymous with unparalleled quality and service to customers and surgical instrumentation produced with the utmost attention to consistent detail. The Miltex instruments includes both premium-grade German and American crafted stainless steel surgical instrumentation manufactured to exacting specifications. The surgical “feel” of these instruments, the outstanding cutting ability and the attention to delicate detail are just a few of the aspects that allow Miltex to outperform and stand out from the competition. These are Miltex Uterine Biopsy Forceps. Uterine Biopsy Forceps are commonly used for biopsy of the uterine cervix in colposcopy. They are made from premium-grade German stainless steel. Specifications: 9in Shaft, 6mm Bite, 30-1480